accessibility menu, dialog, popup

UserName

CAS RN 2246-44-8

IUPAC Name: 2-methylnaphthalen-1-amine
Synonyms: 1-naphthylamine, 2-methyl-
Molecular Weight (g/mol): 157.22
Molecular Formula: C11H11N
InChi Key: JMBLSGAXSMOKPN-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results