missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-methylanthraquinone 90.0+%, TCI America™

Catalog No. A125525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A125525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A125525G Supplier TCI America Supplier No. A125525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82-28-0
Molecular Formula C15H11NO2
Molecular Weight (g/mol) 237.26
MDL Number MFCD00001220
InChI Key ZLCUIOWQYBYEBG-UHFFFAOYSA-N
Synonym Disperse Orange 11
PubChem CID 6702
ChEBI CHEBI:82382
IUPAC Name 1-amino-2-methyl-9,10-dihydroanthracene-9,10-dione
SMILES CC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1N

Specifications

Melting Point 201°C
Quantity 25 g
Formula Weight 237.26
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-2-methylanthraquinone
RTECSNumber CB5740000
TSCA Yes