accessibility menu, dialog, popup

UserName

H3COONH2

CAS RN 82-28-0

IUPAC Name: 1-amino-2-methyl-9,10-dihydroanthracene-9,10-dione
Synonyms: 1-amino-2-methylanthracene-9,10-dione
Molecular Weight (g/mol): 237.26
Molecular Formula: C15H11NO2
InChi Key: ZLCUIOWQYBYEBG-UHFFFAOYSA-N
SMILES: CC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results