missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-11-azido-3,6,9-trioxaundecane, Thermo Scientific Chemicals

Catalog No. p-7042909
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64308MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64308MC Supplier Thermo Scientific Chemicals Supplier No. J64308MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134179-38-7 | Molecular Formula: C8H18N4O3 | Molecular Weight: 218.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134179-38-7
Molecular Formula C8H18N4O3
Molecular Weight (g/mol) 218.257
MDL Number MFCD00269874
InChI Key FPVCVHVTMPCZTH-UHFFFAOYSA-N
Synonym 1-amino-11-azido-3,6,9-trioxaundecane, azido-peg3-amine, 11-azido-3,6,9-trioxaundecan-1-amine, 2-2-2-2-azidoethoxy ethoxy ethoxy ethanamine, 1-2-2-aminoethoxy ethoxy-2-2-azidoethoxy ethane, 11-azido-3 6 9-trioxaundecan-1-amine, ethanamine, 2-2-2-2-azidoethoxy ethoxy ethoxy, 2-2-2-2-azidoethoxy ethoxy ethoxy ethylamine
PubChem CID 2735280
IUPAC Name 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethanamine
SMILES C(COCCOCCOCCN=[N+]=[N-])N

Specifications

Color Brown to Yellow
Linear Formula N3CH2CH2(OCH2CH2)3NH2
Quantity 100 mg
UN Number UN2735
Formula Weight 218.26
Physical Form Viscous Liquid
Chemical Name or Material 1-Amino-11-azido-3,6,9-trioxaundecane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only