missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-1-cyclopropanecarbonitrile hydrochloride, 97%

Catalog No. AAH2728803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2728803 1 g
AAH2728806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2728803 Supplier Thermo Scientific Chemicals Supplier No. H2728803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127946-77-4 | C4H7ClN2 | 118.564 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127946-77-4
Molecular Formula C4H7ClN2
Molecular Weight (g/mol) 118.564
MDL Number MFCD04114063
InChI Key PCEIEQLJYDMRFZ-UHFFFAOYSA-N
Synonym 1-amino-1-cyclopropanecarbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hydrochloride, 1-aminocyclopropane-1-carbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hcl, 1-amino-cyclopropyl cyanic hydrochloride, 1-amino-1-cyanocyclopropane hydrochloride, 1-amino-cyclopropanecarbonitrile hydrochloride, cyclopropanecarbonitrile, 1-amino-, monohydrochloride, 1-cyanocyclopropanaminium chloride, pubchem16832
PubChem CID 14586363
IUPAC Name 1-aminocyclopropane-1-carbonitrile;hydrochloride
SMILES C1CC1(C#N)N.Cl

Specifications

Melting Point ∽220°C (decomposition)
Quantity 1 g
UN Number UN3439
Formula Weight 118.56
Percent Purity 97%
Chemical Name or Material 1-Amino-1-cyclopropanecarbonitrile hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only