missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amidinopyrazole Hydrochloride 98.0+%, TCI America™
$53.90 - $144.39
Chemical Identifiers
| CAS | 4023-02-3 |
|---|---|
| Molecular Formula | C4H6N4 |
| Molecular Weight (g/mol) | 110.12 |
| MDL Number | MFCD00210087 |
| InChI Key | UCQFSGCWHRTMGG-UHFFFAOYSA-N |
| Synonym | 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl |
| PubChem CID | 2734672 |
| IUPAC Name | pyrazole-1-carboximidamide;hydrochloride |
| SMILES | C1=CN(N=C1)C(=N)N.Cl |
Chemical Identifiers
| 4023-02-3 | |
| 110.12 | |
| UCQFSGCWHRTMGG-UHFFFAOYSA-N | |
| 2734672 | |
| C1=CN(N=C1)C(=N)N.Cl |
| C4H6N4 | |
| MFCD00210087 | |
| 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl | |
| pyrazole-1-carboximidamide;hydrochloride |
Specifications
| 4023-02-3 | |
| Yellow | |
| MFCD00210087 | |
| 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl | |
| C1=CN(N=C1)C(=N)N.Cl | |
| 110.12 | |
| 146.58 | |
| Crystalline Powder |
| 167°C | |
| C4H6N4 | |
| 5 g | |
| UCQFSGCWHRTMGG-UHFFFAOYSA-N | |
| pyrazole-1-carboximidamide;hydrochloride | |
| 2734672 | |
| ≥98.0% (HPLC,T) | |
| 1-Amidinopyrazole Hydrochloride |
Safety and Handling
TSCA : No