missing translation for 'onlineSavingsMsg'
Learn More

1-Amidinopyrazole Hydrochloride 98.0+%, TCI America™

Catalog No. a20555g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A20555G 5 g
A205525G 25 g
2 options

Catalog No. A20555G

Supplier: TCI America A20555G

Only null left

Chemical Identifiers

CAS 4023-02-3
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.12
MDL Number MFCD00210087
InChI Key UCQFSGCWHRTMGG-UHFFFAOYSA-N
Synonym 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl
PubChem CID 2734672
IUPAC Name pyrazole-1-carboximidamide;hydrochloride
SMILES C1=CN(N=C1)C(=N)N.Cl

Specifications

Melting Point 167°C
Color Yellow
Quantity 5 g
Formula Weight 146.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Amidinopyrazole Hydrochloride