missing translation for 'onlineSavingsMsg'
Learn More

1-Allyl-3-methylimidazolium bromide, 97%

Catalog No. AAH5951314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5951314 25 g
AAH5951306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5951314 Supplier Thermo Scientific Chemicals Supplier No. H5951314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31410-07-8 | C7H11BrN2 | 203.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31410-07-8
Molecular Formula C7H11BrN2
Molecular Weight (g/mol) 203.08
MDL Number MFCD08277003
InChI Key KLFDZFIZKMEUGI-UHFFFAOYSA-M
Synonym 1-allyl-3-methylimidazolium bromide, 1-allyl-3-methyl-1h-imidazol-3-ium bromide, 1-methyl-3-prop-2-en-1-yl imidazol-1-ium bromide, ksc221i2h, 3-methyl-1-prop-2-en-1-yl imidazol-1-ium bromide
PubChem CID 11229454
SMILES [Br-].C[N+]1=CN(CC=C)C=C1

Specifications

Color Yellow
Quantity 25 g
Sensitivity Moisture sensitive
IUPAC Name 3-methyl-1-(prop-2-en-1-yl)-1H-imidazol-3-ium bromide
Formula Weight 203.08
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Allyl-3-methylimidazolium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only