accessibility menu, dialog, popup

UserName

CAS RN 31410-07-8

IUPAC Name: 3-methyl-1-(prop-2-en-1-yl)-1H-imidazol-3-ium bromide
Synonyms: 1-methyl-3-(prop-2-en-1-yl)imidazol-1-ium bromide
Molecular Weight (g/mol): 203.08
Molecular Formula: C7H11BrN2
InChi Key: KLFDZFIZKMEUGI-UHFFFAOYSA-M
SMILES: [Br-].C[N+]1=CN(CC=C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results