missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Allyl-3-(2-hydroxyethyl)-2-thiourea 98.0+%, TCI America™
$39.14 - $39.14
Chemical Identifiers
| CAS | 105-81-7 |
|---|---|
| Molecular Formula | C6H12N2OS |
| Molecular Weight (g/mol) | 160.235 |
| MDL Number | MFCD00002838 |
| InChI Key | VUVPNTYTOUGMDG-UHFFFAOYSA-N |
| Synonym | 1-allyl-3-2-hydroxyethyl-2-thiourea, 1-allyl-3-2-hydroxyethyl thiourea, thiourea, n-2-hydroxyethyl-n'-2-propenyl, n-allyl-n'-2-hydroxyethyl thiourea, n-2-hydroxyethyl-n'-allylthiourea, n-2-hydroxyethyl-n'-2-propenylthiourea, n-allyl-n'-2-hydroxyethylthiourea, urea, 1-allyl-3-2-hydroxyethyl-2-thio, n-allyl-n'-beta-hydroxyethyl thiourea |
| PubChem CID | 2735273 |
| IUPAC Name | 1-(2-hydroxyethyl)-3-prop-2-enylthiourea |
| SMILES | C=CCNC(=S)NCCO |
Chemical Identifiers
| 105-81-7 | |
| 160.235 | |
| VUVPNTYTOUGMDG-UHFFFAOYSA-N | |
| 2735273 | |
| C=CCNC(=S)NCCO |
| C6H12N2OS | |
| MFCD00002838 | |
| 1-allyl-3-2-hydroxyethyl-2-thiourea, 1-allyl-3-2-hydroxyethyl thiourea, thiourea, n-2-hydroxyethyl-n'-2-propenyl, n-allyl-n'-2-hydroxyethyl thiourea, n-2-hydroxyethyl-n'-allylthiourea, n-2-hydroxyethyl-n'-2-propenylthiourea, n-allyl-n'-2-hydroxyethylthiourea, urea, 1-allyl-3-2-hydroxyethyl-2-thio, n-allyl-n'-beta-hydroxyethyl thiourea | |
| 1-(2-hydroxyethyl)-3-prop-2-enylthiourea |
Specifications
| 105-81-7 | |
| White-Yellow | |
| MFCD00002838 | |
| 1-allyl-3-2-hydroxyethyl-2-thiourea, 1-allyl-3-2-hydroxyethyl thiourea, thiourea, n-2-hydroxyethyl-n'-2-propenyl, n-allyl-n'-2-hydroxyethyl thiourea, n-2-hydroxyethyl-n'-allylthiourea, n-2-hydroxyethyl-n'-2-propenylthiourea, n-allyl-n'-2-hydroxyethylthiourea, urea, 1-allyl-3-2-hydroxyethyl-2-thio, n-allyl-n'-beta-hydroxyethyl thiourea | |
| C=CCNC(=S)NCCO | |
| 160.235 | |
| 160.24 | |
| Crystalline Powder |
| 78°C | |
| C6H12N2OS | |
| 25 g | |
| VUVPNTYTOUGMDG-UHFFFAOYSA-N | |
| 1-(2-hydroxyethyl)-3-prop-2-enylthiourea | |
| 2735273 | |
| ≥98.0% (N) | |
| 1-Allyl-3-(2-hydroxyethyl)-2-thiourea |
Safety and Handling
EINECSNumber : (2)-1764
RTECSNumber : YR7913000
TSCA : Yes