missing translation for 'onlineSavingsMsg'
Learn More

1-Allyl-2-thiourea 98.0+%, TCI America™

Supplier:  TCI America A0220100G

 View more versions of this product

Catalog No. A0220100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Allyl-2-thiourea
71°C
C4H8N2S
100 g
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
C=CCNC(=S)N
116.182
CHEBI:74079
≥98.0% (T)
109-57-9
White-Yellow
MFCD00004940
2811
HTKFORQRBXIQHD-UHFFFAOYSA-N
prop-2-enylthiourea
1549517
116.18
Crystalline Powder

Chemical Identifiers

109-57-9
116.182
HTKFORQRBXIQHD-UHFFFAOYSA-N
1549517
prop-2-enylthiourea
C4H8N2S
MFCD00004940
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
CHEBI:74079
C=CCNC(=S)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YR8050000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions