missing translation for 'onlineSavingsMsg'
Learn More

1-Allyl-2-thiourea 98.0+%, TCI America™

Catalog No. A022025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A022025G 25 g
A0220100G 100 g
A0220500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A022025G Supplier TCI America Supplier No. A022025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-57-9
Molecular Formula C4H8N2S
Molecular Weight (g/mol) 116.182
MDL Number MFCD00004940
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 116.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Allyl-2-thiourea
RTECSNumber YR8050000
TSCA Yes