missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl methyl ketone, 99%, Thermo Scientific™

Chemical Identifiers

CAS 1660-04-4
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00074739
InChI Key DACIGVIOAFXPHW-UHFFFAOYSA-N
Synonym 1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone
PubChem CID 123126
IUPAC Name 1-(1-adamantyl)ethanone
SMILES CC(=O)C12CC3CC(CC(C3)C1)C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC164740010
SDS View Documents
Thermo Scientific Chemicals
164740010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC164740050
SDS View Documents
Thermo Scientific Chemicals
164740050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC164740250
SDS View Documents
Thermo Scientific Chemicals
164740250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1660-04-4
178.28
DACIGVIOAFXPHW-UHFFFAOYSA-N
123126
CC(=O)C12CC3CC(CC(C3)C1)C2
C12H18O
MFCD00074739
1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone
1-(1-adamantyl)ethanone

Specifications

1660-04-4
100.0
Authentic
Glass bottle
MFCD00074739
1-adamantyl methyl ketone, 1-acetyladamantane, 1-1-adamantyl ethanone, 1-adamantylmethylketone, 1-adamantan-1-yl ethan-1-one, 1-adamantan-1-yl ethanone, acetyladamantane, methyl 1-adamantyl ketone, 1-adamantan-1-yl-ethanone, adamantyl methyl ketone
CC(=O)C12CC3CC(CC(C3)C1)C2
178.28
178.27
1-Adamantyl methyl ketone, 99%
98.5
108°C
99%
C12H18O
1g
DACIGVIOAFXPHW-UHFFFAOYSA-N
1-(1-adamantyl)ethanone
123126
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-761-9

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only