missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Adamantanemethylamine 98.0+%, TCI America™
$240.46 - $715.79
Chemical Identifiers
| CAS | 17768-41-1 |
|---|---|
| Molecular Formula | C11H20N |
| Molecular Weight (g/mol) | 166.29 |
| MDL Number | MFCD00074750 |
| InChI Key | XSOHXMFFSKTSIT-UHFFFAOYSA-O |
| Synonym | 1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine |
| PubChem CID | 86625 |
| IUPAC Name | (adamantan-1-yl)methanaminium |
| SMILES | [NH3+]CC12CC3CC(CC(C3)C1)C2 |
Chemical Identifiers
| 17768-41-1 | |
| 166.29 | |
| XSOHXMFFSKTSIT-UHFFFAOYSA-O | |
| 86625 | |
| [NH3+]CC12CC3CC(CC(C3)C1)C2 |
| C11H20N | |
| MFCD00074750 | |
| 1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine | |
| (adamantan-1-yl)methanaminium |
Specifications
| 17768-41-1 | |
| 97°C | |
| MFCD00074750 | |
| 1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine | |
| [NH3+]CC12CC3CC(CC(C3)C1)C2 | |
| 166.29 | |
| 165.28 | |
| Liquid |
| Yellow | |
| C11H20N | |
| 1 g | |
| XSOHXMFFSKTSIT-UHFFFAOYSA-O | |
| (adamantan-1-yl)methanaminium | |
| 86625 | |
| ≥98.0% (T) | |
| 1-Adamantanemethylamine |
Safety and Handling
TSCA : No