missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneethanol 98.0+%, TCI America™

Catalog No. p-7117550
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16195G 5 g
A161925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16195G Supplier TCI America Supplier No. A16195G
Only null left

Chemical Identifiers

CAS 6240-11-5
Molecular Formula C12H20O
Molecular Weight (g/mol) 180.29
MDL Number MFCD00074756
InChI Key ZBIDZPHRNBZTLT-UHFFFAOYSA-N
Synonym 1-adamantaneethanol, 2-adamantan-1-yl ethanol, 2-1-adamantyl ethanol, 1-admantaneethanol, 1-adamantane ethanol, tricyclo 3.3.1.13,7 decaneethanol 9ci, tricyclo 3.3.1.1 3,7-decane-1-ethanol, 2-tricyclo 3.3.1.1'3,7 dec-1-yl ethanol, 2-3r,5s,7s-adamantan-1-yl ethanol, 2-adamantan-1-ylethanol
PubChem CID 110810
IUPAC Name 2-(1-adamantyl)ethanol
SMILES C1C2CC3CC1CC(C2)(C3)CCO

Specifications

Melting Point 76°C
Color White
Quantity 5 g
Formula Weight 180.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantaneethanol