missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneethanol, 98%

Catalog No. AAL0931806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0931806 5 g
AAL0931803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0931806 Supplier Thermo Scientific Chemicals Supplier No. L0931806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6240-11-5 | C12H22O2 | 198.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6240-11-5
MDL Number MFCD00074756

Specifications

Melting Point 73°C to 75°C
Molecular Formula C12H20O
Quantity 5 g
Beilstein 1855382
Synonym 1-adamantaneethanol, 2-adamantan-1-yl ethanol, 2-1-adamantyl ethanol, 1-admantaneethanol, 1-adamantane ethanol, tricyclo 3.3.1.13,7 decaneethanol 9ci, tricyclo 3.3.1.1 3,7-decane-1-ethanol, 2-tricyclo 3.3.1.1'3,7 dec-1-yl ethanol, 2-3r,5s,7s-adamantan-1-yl ethanol, 2-adamantan-1-ylethanol
InChI Key ZBIDZPHRNBZTLT-UHFFFAOYSA-N
SMILES C1C2CC3CC1CC(C2)(C3)CCO
IUPAC Name 2-(1-adamantyl)ethanol
Molecular Weight (g/mol) 180.29
PubChem CID 110810
Formula Weight 180.29
Percent Purity 98%
Chemical Name or Material 1-Adamantaneethanol
Show More Show Less
EINECSNumber 228-353-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only