missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneethanol, 98%

Catalog No. AAL0931803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0931803 1 g
AAL0931806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0931803 Supplier Thermo Scientific Chemicals Supplier No. L0931803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6240-11-5 | C12H22O2 | 198.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6240-11-5
MDL Number MFCD00074756

Specifications

Melting Point 73°C to 75°C
Molecular Formula C12H22O2
Quantity 1 g
Beilstein 1855382
Synonym hexanoic acid, cyclohexyl ester, cyclohexyl caproate, unii-v3b77j8gnh, v3b77j8gnh, cyclohexyl n-hexanoate, hexanoic acid cyclohexyl ester, acmc-1b676, 4-4-methoxyphenyl-1,3-thiazol-4-amine
InChI Key DTNOERNOMHQUCN-UHFFFAOYSA-N
SMILES CCCCCC(=O)OC1CCCCC1
IUPAC Name cyclohexyl hexanoate
Molecular Weight (g/mol) 198.31
PubChem CID 80388
Formula Weight 180.29
Percent Purity 98%
Chemical Name or Material 1-Adamantaneethanol
Show More Show Less
EINECSNumber 228-353-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only