missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanecarboxylic acid chloride, 97%

Catalog No. p-50413
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152480050 5 g Glass bottle
AC152480250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152480050 Supplier Thermo Scientific Chemicals Supplier No. 152480050
Only null left

CAS: 2094-72-6 | C11H15ClO | 198.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2094-72-6
Molecular Formula C11H15ClO
Molecular Weight (g/mol) 198.69
MDL Number MFCD00074724
InChI Key MIBQYWIOHFTKHD-UHFFFAOYSA-N
Synonym 1-adamantanecarbonyl chloride, adamantanecarbonyl chloride, 1-adamantanecarbonylchloride, 1-adamantoyl chloride, adamantoyl chloride, 1-adamantanoic acid chloride, 1-adamantanecarboxylic acid chloride, adamantane-1-carboxylic acid chloride, 1-adamantylcarbonyl chloride, 1-adamantyl carbonyl chloride
PubChem CID 98915
IUPAC Name adamantane-1-carbonyl chloride
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 48.0°C to 52.0°C
Color White
Boiling Point 135.0°C to 136.0°C (10.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Fieser 02,15
Solubility Information Solubility in water: may decompose
Formula Weight 198.69
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 1-Adamantanecarboxylic acid chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-252-7
TSCA TSCA

RUO – Research Use Only