missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanecarbonyl Chloride 97.0+%, TCI America™

Catalog No. p-7117022
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07175G 5 g
A071725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07175G Supplier TCI America Supplier No. A07175G
Only null left

Chemical Identifiers

CAS 2094-72-6
Molecular Formula C11H15ClO
Molecular Weight (g/mol) 198.69
MDL Number MFCD00074724
InChI Key MIBQYWIOHFTKHD-UHFFFAOYSA-N
Synonym 1-adamantanecarbonyl chloride, adamantanecarbonyl chloride, 1-adamantanecarbonylchloride, 1-adamantoyl chloride, adamantoyl chloride, 1-adamantanoic acid chloride, 1-adamantanecarboxylic acid chloride, adamantane-1-carboxylic acid chloride, 1-adamantylcarbonyl chloride, 1-adamantyl carbonyl chloride
PubChem CID 98915
IUPAC Name adamantane-1-carbonyl chloride
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)Cl

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 136°C
Quantity 5 g
UN Number 3261
Formula Weight 198.69
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanecarbonyl Chloride