missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 665-66-7 |
|---|---|
| Molecular Formula | C10H18ClN |
| Molecular Weight (g/mol) | 187.71 |
| MDL Number | MFCD00074723 |
| InChI Key | WOLHOYHSEKDWQH-UHFFFAOYSA-N |
| Synonym | amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane |
| PubChem CID | 64150 |
| ChEBI | CHEBI:2619 |
| IUPAC Name | adamantan-1-amine;hydrochloride |
| SMILES | [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC151670250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.
Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol
Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
Chemical Identifiers
| C10H18ClN | |
| MFCD00074723 | |
| amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane | |
| CHEBI:2619 | |
| [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2 |
Specifications
| 665-66-7 | |
| 100 | |
| 99+% | |
| C10H18ClN | |
| 15, 368 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether | |
| [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2 | |
| 187.71 | |
| CHEBI:2619 | |
| 99+% |
| 99 | |
| Authentic | |
| Glass bottle | |
| MFCD00074723 | |
| amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane | |
| WOLHOYHSEKDWQH-UHFFFAOYSA-N | |
| adamantan-1-amine;hydrochloride | |
| 64150 | |
| 187.71 | |
| 1-Adamantanamine hydrochloride, 99+% |
Safety and Handling
Harmful if swallowed
Acute toxicity (category 4)
EINECSNumber : 211-560-2
RTECSNumber : AU4375000
TSCA : TSCA
RUO – Research Use Only