missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Acryloyloxy)-3-(methacryloyloxy)-2-propanol (stabilized with MEHQ) 80.0+%, TCI America™
$37.29 - $156.59
Chemical Identifiers
| CAS | 1709-71-3 |
|---|---|
| Molecular Formula | C10H14O5 |
| Molecular Weight (g/mol) | 214.217 |
| MDL Number | MFCD00274303 |
| InChI Key | ZODNDDPVCIAZIQ-UHFFFAOYSA-N |
| Synonym | 3-(Acryloyloxy)-2-hydroxypropyl Methacrylate |
| PubChem CID | 5062351 |
| IUPAC Name | (2-hydroxy-3-prop-2-enoyloxypropyl) 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OCC(COC(=O)C=C)O |
Chemical Identifiers
| 1709-71-3 | |
| 214.217 | |
| ZODNDDPVCIAZIQ-UHFFFAOYSA-N | |
| 5062351 | |
| CC(=C)C(=O)OCC(COC(=O)C=C)O |
| C10H14O5 | |
| MFCD00274303 | |
| 3-(Acryloyloxy)-2-hydroxypropyl Methacrylate | |
| (2-hydroxy-3-prop-2-enoyloxypropyl) 2-methylprop-2-enoate |
Specifications
| 1709-71-3 | |
| C10H14O5 | |
| 25 g | |
| ZODNDDPVCIAZIQ-UHFFFAOYSA-N | |
| (2-hydroxy-3-prop-2-enoyloxypropyl) 2-methylprop-2-enoate | |
| 5062351 | |
| ≥80.0% (GC) | |
| 1-(Acryloyloxy)-3-(methacryloyloxy)-2-propanol (stabilized with MEHQ) |
| Yellow | |
| MFCD00274303 | |
| 3-(Acryloyloxy)-2-hydroxypropyl Methacrylate | |
| CC(=C)C(=O)OCC(COC(=O)C=C)O | |
| 214.217 | |
| 214.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0958
TSCA : No