missing translation for 'onlineSavingsMsg'
Learn More

1-(Acryloyloxy)-3-(methacryloyloxy)-2-propanol (stabilized with MEHQ) 80.0+%, TCI America™

Catalog No. A198825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A198825G 25 g
A1988500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A198825G Supplier TCI America Supplier No. A198825G
Only null left

Chemical Identifiers

CAS 1709-71-3
Molecular Formula C10H14O5
Molecular Weight (g/mol) 214.217
MDL Number MFCD00274303
InChI Key ZODNDDPVCIAZIQ-UHFFFAOYSA-N
Synonym 3-(Acryloyloxy)-2-hydroxypropyl Methacrylate
PubChem CID 5062351
IUPAC Name (2-hydroxy-3-prop-2-enoyloxypropyl) 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(COC(=O)C=C)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Acryloyloxy)-3-(methacryloyloxy)-2-propanol (stabilized with MEHQ)
EINECSNumber (2)-0958
TSCA No