missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperidine-4-carboxylic acid, 98+%

CAS: 25503-90-6 | C8H12NO3 | 170.19 g/mol

Supplier:  Thermo Scientific Chemicals A1560614

 View more versions of this product

Catalog No. AAA1560614


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Acetylpiperidine-4-carboxylic acid
180°C to 184°C
MFCD00023706
389577
WFCLWJHOKCQYOQ-UHFFFAOYSA-M
1-acetylpiperidine-4-carboxylic acid
117255
≥98%
25503-90-6
C8H12NO3
25 g
1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl
CC(=O)N1CCC(CC1)C([O-])=O
170.19
171.19

Chemical Identifiers

25503-90-6
170.19
WFCLWJHOKCQYOQ-UHFFFAOYSA-M
117255
CC(=O)N1CCC(CC1)C([O-])=O
C8H12NO3
MFCD00023706
1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl
1-acetylpiperidine-4-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H335

EINECSNumber : 247-049-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only