missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylnaphthalene, 97+%

Catalog No. p-7020833
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1291818 50 g
AAA1291830 250 g
AAA129180B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1291818 Supplier Thermo Scientific Chemicals Supplier No. A1291818
Only null left

CAS: 941-98-0 | C12H10O | 170.211 g/mol

They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 941-98-0
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00004013
InChI Key QQLIGMASAVJVON-UHFFFAOYSA-N
Synonym 1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone
PubChem CID 13663
IUPAC Name 1-naphthalen-1-ylethanone
SMILES CC(=O)C1=CC=CC2=CC=CC=C21

Specifications

Melting Point 9°C to 11°C
Density 1.12
Boiling Point 302°C
Flash Point >110°C (230°F)
Refractive Index 1.628
Quantity 50 g
Beilstein 1100618
Solubility Information Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil.
Formula Weight 170.21
Percent Purity ≥97%
Chemical Name or Material 1-Acetylnaphthalene
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-384-1
RTECSNumber AL2982800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only