missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylimidazole, 98%, Thermo Scientific™

Chemical Identifiers

CAS 2466-76-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00005287
InChI Key VIHYIVKEECZGOU-UHFFFAOYSA-N
Synonym 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
PubChem CID 17174
ChEBI CHEBI:16984
IUPAC Name 1-imidazol-1-ylethanone
SMILES CC(=O)N1C=CN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169500250
SDS View Documents
Thermo Scientific Chemicals
169500250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC169501000
SDS View Documents
Thermo Scientific Chemicals
169501000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2466-76-4
110.12
VIHYIVKEECZGOU-UHFFFAOYSA-N
17174
1-imidazol-1-ylethanone
C5H6N2O
MFCD00005287
1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
CHEBI:16984
CC(=O)N1C=CN=C1

Specifications

2466-76-4
100.0
98%
C5H6N2O
25g
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ether,chloroform,thf
CC(=O)N1C=CN=C1
110.12
CHEBI:16984
98%
97.5
Authentic
Glass bottle
MFCD00005287
1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
VIHYIVKEECZGOU-UHFFFAOYSA-N
1-imidazol-1-ylethanone
17174
110.12
1-Acetylimidazole, 98%
Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

EINECSNumber : 219-577-7

RTECSNumber : NI3400000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only