missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetylimidazole, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 169500250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 1-Acetylimidazole | |
| 2466-76-4 | |
| 100.0 | |
| 98% | |
| C5H6N2O | |
| 25g | |
| Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ether,chloroform,thf | |
| CC(=O)N1C=CN=C1 | |
| 110.12 | |
| CHEBI:16984 | |
| 98% |
| 98% | |
| 97.5 | |
| Authentic | |
| Glass bottle | |
| MFCD00005287 | |
| 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534 | |
| VIHYIVKEECZGOU-UHFFFAOYSA-N | |
| 1-imidazol-1-ylethanone | |
| 17174 | |
| 110.12 |
Chemical Identifiers
| 2466-76-4 | |
| 110.12 | |
| VIHYIVKEECZGOU-UHFFFAOYSA-N | |
| 17174 | |
| 1-imidazol-1-ylethanone |
| C5H6N2O | |
| MFCD00005287 | |
| 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534 | |
| CHEBI:16984 | |
| CC(=O)N1C=CN=C1 |
Safety and Handling
MOISTURE SENSITIVE
EINECSNumber : 219-577-7
RTECSNumber : NI3400000
TSCA : TSCA
RUO â Research Use Only