missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-5-bromoindoline 98.0+%, TCI America™

Catalog No. p-7118149
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29245G 5 g
A292425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29245G Supplier TCI America Supplier No. A29245G

Chemical Identifiers

CAS 22190-38-1
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD00056017
InChI Key WQKQAIXOTCPWFE-UHFFFAOYSA-N
Synonym 1-acetyl-5-bromoindoline, 1-5-bromoindolin-1-yl ethanone, 1-5-bromo-2,3-dihydro-1h-indol-1-yl ethan-1-one, 1-5-bromo-2,3-dihydroindol-1-yl ethanone, 1-5-bromo-2,3-dihydro-1h-indol-1-yl ethanone, 1h-indole, 1-acetyl-5-bromo-2,3-dihydro, 1-acetyl-5-bromo-2,3-dihydroindole, acmc-20ailu, 5-bromo-n-acetylindoline, maybridge1_005011
PubChem CID 721847
IUPAC Name 1-(5-bromo-2,3-dihydroindol-1-yl)ethanone
SMILES CC(=O)N1CCC2=C1C=CC(=C2)Br

Specifications

Melting Point 121°C
Quantity 5 g
Formula Weight 240.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetyl-5-bromoindoline
TSCA No
Recommended Storage Freezer