accessibility menu, dialog, popup

UserName

CAS RN 22190-38-1

H3CONBr

CAS RN 22190-38-1

IUPAC Name: 1-(5-bromo-2,3-dihydro-1H-indol-1-yl)ethan-1-one
Synonyms: 1-(5-bromo-2,3-dihydroindol-1-yl)ethanone
Molecular Weight (g/mol): 240.1
Molecular Formula: C10H10BrNO
InChi Key: WQKQAIXOTCPWFE-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC2=CC(Br)=CC=C12
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results