missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-5-bromoindoline, 98%

Catalog No. AAB2050914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2050914 25 g
AAB2050903 1 g
AAB2050906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2050914 Supplier Thermo Scientific Chemicals Supplier No. B2050914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22190-38-1 | C10H10BrNO | 240.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22190-38-1
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD00056017
InChI Key WQKQAIXOTCPWFE-UHFFFAOYSA-N
Synonym 1-acetyl-5-bromoindoline, 1-5-bromoindolin-1-yl ethanone, 1-5-bromo-2,3-dihydro-1h-indol-1-yl ethan-1-one, 1-5-bromo-2,3-dihydroindol-1-yl ethanone, 1-5-bromo-2,3-dihydro-1h-indol-1-yl ethanone, 1h-indole, 1-acetyl-5-bromo-2,3-dihydro, 1-acetyl-5-bromo-2,3-dihydroindole, acmc-20ailu, 5-bromo-n-acetylindoline, maybridge1_005011
PubChem CID 721847
IUPAC Name 1-(5-bromo-2,3-dihydroindol-1-yl)ethanone
SMILES CC(=O)N1CCC2=C1C=CC(=C2)Br

Specifications

Melting Point 118°C to 121°C
Quantity 25 g
Beilstein 165052
Formula Weight 240.1
Percent Purity 98%
Chemical Name or Material 1-Acetyl-5-bromoindoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only