missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-5-bromo-4-chloro-3-indolyl Acetate 98.0+%, TCI America™

Catalog No. p-7118148
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29231G 1 g
A29235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29231G Supplier TCI America Supplier No. A29231G

Chemical Identifiers

CAS 3030-06-6
Molecular Formula C12H9BrClNO3
Molecular Weight (g/mol) 330.56
MDL Number MFCD00040632
InChI Key DSHQTSIXXYZXGR-UHFFFAOYSA-N
Synonym 1-acetyl-5-bromo-4-chloro-1h-indol-3-yl acetate, 5-bromo-4-chloroindoxyl 1,3-diacetate, 5-bromo-4-chloroindoxyl-1,3-diacetate, 3-acetoxy-1-acetyl-5-bromo-4-chloroindole, 1-acetyl-5-bromo-4-chloro-1h-indol-3-ylacetate, 1h-indol-3-ol, 1-acetyl-5-bromo-4-chloro-, acetate ester, acmc-1ag9t, 5-bromo-4-chloroindolyl 1,3-diacetate, 5-bromo-4-chloro-indoxyl-1,3-diacetate, cas
PubChem CID 76416
IUPAC Name (1-acetyl-5-bromo-4-chloroindol-3-yl) acetate
SMILES CC(=O)N1C=C(C2=C1C=CC(=C2Cl)Br)OC(=O)C

Specifications

Melting Point 166°C
Quantity 1 g
Formula Weight 330.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetyl-5-bromo-4-chloro-3-indolyl Acetate
TSCA No
Recommended Storage Refrigerator