accessibility menu, dialog, popup

UserName

CAS RN 3030-06-6

IUPAC Name: 1-acetyl-5-bromo-4-chloro-1H-indol-3-yl acetate
Synonyms: 1-acetyl-5-bromo-4-chloroindol-3-yl acetate
Molecular Weight (g/mol): 330.56
Molecular Formula: C12H9BrClNO3
InChi Key: DSHQTSIXXYZXGR-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CN(C(C)=O)C2=CC=C(Br)C(Cl)=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results