missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-fluoronaphthalene, 97%

Catalog No. p-7027934
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2500403 1 g
AAB2500414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2500403 Supplier Thermo Scientific Chemicals Supplier No. B2500403
Only null left

CAS: 316-68-7 | C12H9FO | 188.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 316-68-7
Molecular Formula C12H9FO
Molecular Weight (g/mol) 188.201
MDL Number MFCD00134475
InChI Key GAMOBQXCYKWQLT-UHFFFAOYSA-N
Synonym 1-4-fluoronaphthalen-1-yl ethanone, 1-acetyl-4-fluoronaphthalene, 1-4-fluoro-1-naphthyl ethanone, 4-fluoro-1-acetonaphthone, ethanone, 1-4-fluoro-1-naphthalenyl, 1-4-fluoro-1-naphthyl ethan-1-one, 4'-fluoro-1'-acetonaphthone, 1-4-fluoronaphthalen-1-yl ethan-1-one, 4-fluoroacetonaphthone, 4'-fluoro-1'acetonaphthone
PubChem CID 67568
IUPAC Name 1-(4-fluoronaphthalen-1-yl)ethanone
SMILES CC(=O)C1=CC=C(C2=CC=CC=C21)F

Specifications

Density 1.21
Boiling Point 164°C to 166°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.608
Quantity 1 g
Formula Weight 188.2
Percent Purity 97%
Chemical Name or Material 1-Acetyl-4-fluoronaphthalene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-261-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only