missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine, 96%

Catalog No. p-7031838
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33058MD 250 mg
AAH3305803 1 g
AAH3305806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH33058MD Supplier Thermo Scientific Chemicals Supplier No. H33058MD
Only null left

CAS: 84162-82-3 | C14H15F2NO2 | 267.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84162-82-3
Molecular Formula C14H15F2NO2
Molecular Weight (g/mol) 267.276
MDL Number MFCD00179440
InChI Key UGPWQEIGJWVJDR-UHFFFAOYSA-N
Synonym 4-2',4'-difluorobenzoyl-1-acetylpiperidine, 1-4-2,4-difluorobenzoyl piperidin-1-yl ethanone, 1-acetylpiperidin-4-yl 2,4-difluorophenyl methanone, 1-acetyl-4-2,4-difluorobenzoyl piperidine, 1-4-2,4-difluoro-benzoyl-piperidin-1-yl-ethanone, 1-acetyl-4-2',4'-difluorobenzoyl piperidine, 1-4-2,4-difluorobenzoyl-1-piperidinyl-1-ethanone, 1-4-2.4-difluoro-benzoyl-piperidin-1-yl-ethanone, maybridge3_006593
PubChem CID 2740707
IUPAC Name 1-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethanone
SMILES CC(=O)N1CCC(CC1)C(=O)C2=C(C=C(C=C2)F)F

Specifications

Melting Point 97°C to 98°C
Quantity 250 mg
Formula Weight 267.28
Percent Purity 96%
Chemical Name or Material 1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only