accessibility menu, dialog, popup

UserName

H3CONOFF

CAS RN 84162-82-3

IUPAC Name: 1-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethan-1-one
Synonyms: 1-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethanone
Molecular Weight (g/mol): 267.28
Molecular Formula: C14H15F2NO2
InChi Key: UGPWQEIGJWVJDR-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC(CC1)C(=O)C1=C(F)C=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results