missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetyl-2-imidazolidinone 98.0+%, TCI America™
$76.43 - $378.40
Chemical Identifiers
| CAS | 5391-39-9 |
|---|---|
| Molecular Formula | C5H8N2O2 |
| Molecular Weight (g/mol) | 128.131 |
| MDL Number | MFCD00988254 |
| InChI Key | JJWACYUTERPMBM-UHFFFAOYSA-N |
| PubChem CID | 79350 |
| IUPAC Name | 1-acetylimidazolidin-2-one |
| SMILES | CC(=O)N1CCNC1=O |
Chemical Identifiers
| 5391-39-9 | |
| 128.131 | |
| JJWACYUTERPMBM-UHFFFAOYSA-N | |
| 1-acetylimidazolidin-2-one |
| C5H8N2O2 | |
| MFCD00988254 | |
| 79350 | |
| CC(=O)N1CCNC1=O |
Specifications
| 5391-39-9 | |
| White-Yellow | |
| MFCD00988254 | |
| JJWACYUTERPMBM-UHFFFAOYSA-N | |
| 1-acetylimidazolidin-2-one | |
| 79350 | |
| ≥98.0% (GC) | |
| 1-Acetyl-2-imidazolidinone |
| 185°C | |
| C5H8N2O2 | |
| 5 g | |
| CC(=O)N1CCNC1=O | |
| 128.131 | |
| 128.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No