accessibility menu, dialog, popup

UserName

CAS RN 5391-39-9

H3CONHNO

CAS RN 5391-39-9

IUPAC Name: 1-acetylimidazolidin-2-one
Synonyms: 1-acetylimidazolidin-2-one
Molecular Weight (g/mol): 128.13
Molecular Formula: C5H8N2O2
InChi Key: JJWACYUTERPMBM-UHFFFAOYSA-N
SMILES: CC(=O)N1CCNC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results