missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine 99.0+%, TCI America™

Catalog No. A12191G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12191G Supplier TCI America Supplier No. A12191G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107866-54-6
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.152
MDL Number MFCD00075383
InChI Key SDYATKOQVBKTLQ-UHFFFAOYSA-N
PubChem CID 702889
IUPAC Name 1-(triazolo[4,5-b]pyridin-1-yl)ethanone
SMILES CC(=O)N1C2=C(N=CC=C2)N=N1

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
Formula Weight 162.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine