accessibility menu, dialog, popup

UserName

CAS RN 107866-54-6

IUPAC Name: 1-{1H-[1,2,3]triazolo[4,5-b]pyridin-1-yl}ethan-1-one
Synonyms: 1-{[1,2,3]triazolo[4,5-b]pyridin-1-yl}ethanone
Molecular Weight (g/mol): 162.15
Molecular Formula: C7H6N4O
InChi Key: SDYATKOQVBKTLQ-UHFFFAOYSA-N
SMILES: CC(=O)N1N=NC2=NC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results