missing translation for 'onlineSavingsMsg'
Learn More

1-Acetoxyindan 98.0+%, TCI America™

Catalog No. A19755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19755G Supplier TCI America Supplier No. A19755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26452-98-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00053467
InChI Key BZZWYQKRGRYJAX-UHFFFAOYNA-N
PubChem CID 233153
IUPAC Name 2,3-dihydro-1H-inden-1-yl acetate
SMILES CC(=O)OC1CCC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 241°C
Quantity 5 g
Formula Weight 176.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Acetoxyindan