missing translation for 'onlineSavingsMsg'
Learn More

1-Acetoxy-3-chloroacetone 95.0+%, TCI America™

Catalog No. p-7117321
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12325G 5 g
A123225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12325G Supplier TCI America Supplier No. A12325G

Chemical Identifiers

CAS 40235-68-5
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00191365
InChI Key LCMPIWIWBGHBOY-UHFFFAOYSA-N
Synonym 1-Acetoxy-3-acetenyl Chloride, 1-Acetoxy-3-chloro-2-propanone
PubChem CID 38413
IUPAC Name 3-chloro-2-oxopropyl acetate
SMILES CC(=O)OCC(=O)CCl

Specifications

Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 150.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Acetoxy-3-chloroacetone
RTECSNumber AF8600000
TSCA Yes