missing translation for 'onlineSavingsMsg'
Learn More

1-Acetoxy-2-methoxybenzene, 98%

Catalog No. AAB2024214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2024214 25 g
AAB2024222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2024214 Supplier Thermo Scientific Chemicals Supplier No. B2024214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15212-03-0 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15212-03-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00017221
InChI Key BHJHPYFAYGAPLS-UHFFFAOYSA-N
Synonym guaiacol acetate, 1-acetoxy-2-methoxybenzene, guaiacyl acetate, o-acetoxyanisole, o-methoxyphenyl acetate, o-anisyl acetate, acetyl guaiacol, phenol, 2-methoxy-, acetate, o-acetylguaiacol, eucol
PubChem CID 61155
ChEBI CHEBI:86645
IUPAC Name (2-methoxyphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1OC

Specifications

Quantity 25 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 1-Acetoxy-2-methoxybenzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only