missing translation for 'onlineSavingsMsg'
Learn More

1'-Acetonaphthone 95.0+%, TCI America™

Catalog No. A104425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A104425G 25 g
A1044500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A104425G Supplier TCI America Supplier No. A104425G

Chemical Identifiers

CAS 941-98-0
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00004013
InChI Key QQLIGMASAVJVON-UHFFFAOYSA-N
Synonym 1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone
PubChem CID 13663
IUPAC Name 1-naphthalen-1-ylethanone
SMILES CC(=O)C1=CC=CC2=CC=CC=C21

Specifications

Boiling Point 298°C
Quantity 25 g
Formula Weight 170.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1′-Acetonaphthone
EINECSNumber (4)-0394
RTECSNumber AL2982800
TSCA Yes