missing translation for 'onlineSavingsMsg'
Learn More

1-Acetamidonaphthalene 99.0+%, TCI America™

Catalog No. N005025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N005025G 25 g
N0050250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N005025G Supplier TCI America Supplier No. N005025G

Chemical Identifiers

CAS 575-36-0
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.23
MDL Number MFCD00059128
InChI Key OKQIEBVRUGLWOR-UHFFFAOYSA-N
Synonym 1-acetamidonaphthalene, acetamide, n-1-naphthalenyl, n-1-naphthylacetamide, n-naphthalen-1-yl acetamide, n-1-naphthyl acetamide, n-acetyl-1-naphthylamine, n-acetyl-1-aminonaphthalene, n-naphthylacetamide, acetamide, n-1-naphthyl
PubChem CID 68461
IUPAC Name N-(naphthalen-1-yl)acetamide
SMILES CC(=O)NC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 159°C
Color White
Quantity 25 g
Formula Weight 185.23
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetamidonaphthalene