accessibility menu, dialog, popup

UserName

CH3ONH

CAS RN 575-36-0

IUPAC Name: N-(naphthalen-1-yl)acetamide
Synonyms: N-(naphthalen-1-yl)acetamide
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: OKQIEBVRUGLWOR-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results