missing translation for 'onlineSavingsMsg'
Learn More

1-Acetamidoadamantane, 97%

CAS: 880-52-4 | C12H19NO | 193.29 g/mol

$100.96 - $100.96

Chemical Identifiers

CAS 880-52-4
Molecular Formula C12H19NO
Molecular Weight (g/mol) 193.29
MDL Number MFCD00074730
InChI Key BCVXYGJCDZPKGV-UHFFFAOYSA-N
Synonym 1-acetamidoadamantane, n-1-adamantyl acetamide, n-adamantan-1-yl acetamide, 1-acetylaminoadamantane, 1-adamantylacetamide, acetamide, n-1-adamantyl, n-adamantylacetamide, 1-acetamino adamantane, unii-5283y1voii, n-acetyl adamantamine
PubChem CID 64153
IUPAC Name N-(1-adamantyl)acetamide
SMILES CC(=O)NC12CC3CC(C1)CC(C3)C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC133600250
SDS View Documents
Thermo Scientific Chemicals
133600250
25 g Glass bottle
Each for $100.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

880-52-4
193.29
BCVXYGJCDZPKGV-UHFFFAOYSA-N
64153
CC(=O)NC12CC3CC(C1)CC(C3)C2
C12H19NO
MFCD00074730
1-acetamidoadamantane, n-1-adamantyl acetamide, n-adamantan-1-yl acetamide, 1-acetylaminoadamantane, 1-adamantylacetamide, acetamide, n-1-adamantyl, n-adamantylacetamide, 1-acetamino adamantane, unii-5283y1voii, n-acetyl adamantamine
N-(1-adamantyl)acetamide

Specifications

880-52-4
100.0
White to Yellow
99% min. (GC)
C12H19NO
25 g
1-acetamidoadamantane, n-1-adamantyl acetamide, n-adamantan-1-yl acetamide, 1-acetylaminoadamantane, 1-adamantylacetamide, acetamide, n-1-adamantyl, n-adamantylacetamide, 1-acetamino adamantane, unii-5283y1voii, n-acetyl adamantamine
CC(=O)NC12CC3CC(C1)CC(C3)C2
193.29
193.29
Crystalline Powder
99.0
148.0°C to 150.0°C
Authentic
Glass bottle
MFCD00074730
12, 220
BCVXYGJCDZPKGV-UHFFFAOYSA-N
N-(1-adamantyl)acetamide
64153
99+%
1-Acetamidoadamantane, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-914-9

RTECSNumber : AB4308000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only