accessibility menu, dialog, popup

UserName

CH3ONH

CAS RN 880-52-4

IUPAC Name: N-(adamantan-1-yl)acetamide
Synonyms: N-(adamantan-1-yl)acetamide
Molecular Weight (g/mol): 193.29
Molecular Formula: C12H19NO
InChi Key: BCVXYGJCDZPKGV-UHFFFAOYSA-N
SMILES: CC(=O)NC12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)