missing translation for 'onlineSavingsMsg'
Learn More

1-Acetamidoadamantane, 98%

Catalog No. AAA1722409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1722409 10 g
AAA1722418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1722409 Supplier Thermo Scientific Chemicals Supplier No. A1722409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 880-52-4 | C12H19NO | 193.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 880-52-4
Molecular Formula C12H19NO
Molecular Weight (g/mol) 193.29
MDL Number MFCD00074730
InChI Key BCVXYGJCDZPKGV-UHFFFAOYSA-N
Synonym 1-acetamidoadamantane, n-1-adamantyl acetamide, n-adamantan-1-yl acetamide, 1-acetylaminoadamantane, 1-adamantylacetamide, acetamide, n-1-adamantyl, n-adamantylacetamide, 1-acetamino adamantane, unii-5283y1voii, n-acetyl adamantamine
PubChem CID 64153
IUPAC Name N-(1-adamantyl)acetamide
SMILES CC(=O)NC12CC3CC(C1)CC(C3)C2

Specifications

Melting Point 146°C to 149°C
Quantity 10 g
Formula Weight 193.29
Percent Purity 98%
Chemical Name or Material 1-Acetamidoadamantane
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-914-9
RTECSNumber AB4308000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only