missing translation for 'onlineSavingsMsg'
Learn More

1-Acetamido-4-hydroxyanthraquinone, TCI America™

Catalog No. A11301G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A11301G Supplier TCI America Supplier No. A11301G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7323-62-8
Molecular Formula C16H11NO4
Molecular Weight (g/mol) 281.267
MDL Number MFCD00142576
InChI Key VPZXLTVIBKOBRG-UHFFFAOYSA-N
PubChem CID 146027
IUPAC Name N-(4-hydroxy-9,10-dioxoanthracen-1-yl)acetamide
SMILES CC(=O)NC1=C2C(=C(C=C1)O)C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 198°C
Quantity 1 g
Formula Weight 281.27
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetamido-4-hydroxyanthraquinone