accessibility menu, dialog, popup

UserName

CAS RN 7323-62-8

IUPAC Name: N-(4-hydroxy-9,10-dioxo-9,10-dihydroanthracen-1-yl)acetamide
Synonyms: N-(4-hydroxy-9,10-dioxoanthracen-1-yl)acetamide
Molecular Weight (g/mol): 281.27
Molecular Formula: C16H11NO4
InChi Key: VPZXLTVIBKOBRG-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(O)C2=C1C(=O)C1=CC=CC=C1C2=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results