missing translation for 'onlineSavingsMsg'
Learn More

1-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]benzotriazole 98.0+%, TCI America™

Catalog No. F08715G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08715G Supplier TCI America Supplier No. F08715G
Only null left

Chemical Identifiers

CAS 82911-71-5
Molecular Formula C21H15N3O3
Molecular Weight (g/mol) 357.369
MDL Number MFCD00074939
InChI Key KNSPZTVPSAKHBN-UHFFFAOYSA-N
Synonym fmoc-obt, 9h-fluoren-9-yl methyl 1h-benzo d 1,2,3 triazol-1-yl carbonate, 9-fluorenylmethyl 1-benzotriazolyl carbonate, benzotriazol-1-yl 9-fluorenylmethyl carbonate, 1,2,3-benzotriazol-1-yl 9h-fluoren-9-ylmethyl carbonate, 1-fmoc-oxy benzotriazole, 1-fmoc-oxy-1h-benzotriazole, 9-fluorenylmethyl benzotriazolycarbonate, 9-fluorenylmethylbenzotriazolylcarbonate, 9-fluorenylmethyl 1-benzotriazolyl
PubChem CID 5236567
IUPAC Name benzotriazol-1-yl 9H-fluoren-9-ylmethyl carbonate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)ON4C5=CC=CC=C5N=N4

Specifications

Melting Point 178°C
Color White
Quantity 5 g
Formula Weight 357.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]benzotriazole
TSCA No
Recommended Storage Refrigerator